Learn More
Brady™ Bradylite™ Reflective Numbers and Letters
Bradylite™ Reflective Numbers and Letters are made of B-997, a reflective sheeting backed with a pressure sensitive adhesive.
Supplier:  Brady™ 5920S
 View more versions of this product
Catalog No. 19-807-123
Only null left
Explore available promotions
Description
Description
  • This B-997 material is used for utility plant markings applied outdoors in cold climates
  • Specifically designed to be applied at cold temperatures and has excellent weatherability
  • Service temperature range of -40 to 200° F
  • Average outdoor durability of 4-6 years
Specifications
Specifications
RoHS
Acrylic, Pressure-sensitive
Gloss
Reflective Sheeting
Non-sterile
Ideal for low temperature application
4 to 6 Years
-40°F to 200°F
10 lb./in.
1.5 x 1 in.
25.4 mm
3.8 cm
Engineering
Black, Yellow
Label
Reflective Label
S
75 Gardner Units
25/Pk.
-40°C to +93°C
0.007 in.
1 in.
1.5 in.
Product Suggestions
Product Suggestions