Learn More
Brady™ BBP™ High Temperature Metallized Polyester Tape
BBP™ High Temperature Metallized Polyester Tape B-565 is a metallized polyester film with a permanent acrylic pressure sensitive adhesive and a topcoat specifically formulated for thermal transfer printing.
Supplier:  Brady™ B30C1125565SL
 View more versions of this product
Catalog No. 19211206
Description
Description
  • The B-565 material is designed for identification labels or rating and serial plates that require nameplate-like quality
  • Has an average outdoor durability of 3-5 years
  • Service temperature range of -40 to 230° F
  • Standard adhesive for smooth surfaces
Specifications
Chemical Identifiers
Metallised Polyester Tape for BBP3x Printers
Silver
Tape
Non-sterile
BBP30, BBP31, BBP33, BBP35, and BBP37 printers
1
1 Roll
-40 to 230°F
0.0028 in.
100 ft.
Acrylic, Permanent, Pressure-sensitive
Matte
Polyester
For rating and serial plates that utilize barcodes, alphanumeric, graphic symbols, logos
60 to 101 Gardner Units
3 to 5 Years
Brady Workstation, LabelMark, MarkWare
-40°C to +110°C
0.071 mm
30.48 m
Specifications
Metallised Polyester Tape for BBP3x Printers
Silver
Tape
Non-sterile
BBP30, BBP31, BBP33, BBP35, and BBP37 printers
1
1 Roll
-40 to 230°F
0.0028 in.
100 ft.
1.12 in.
B30C-1125-565-SL
Acrylic, Permanent, Pressure-sensitive
Matte
Polyester
For rating and serial plates that utilize barcodes, alphanumeric, graphic symbols, logos
60 to 101 Gardner Units
3 to 5 Years
Brady Workstation, LabelMark, MarkWare
-40°C to +110°C
0.071 mm
30.48 m
29 mm
Product Suggestions
Product Suggestions