Learn More
Brady™ BBP™ DANGER Pre-Printed Pre-Cut Blank Sign Headers
BBP™ DANGER Pre-Printed Pre-Cut Blank Sign Headers are made of durable, low-shrink vinyl with ultra-aggressive adhesive that is ideal for labeling pipes, equipment, shelves and nearly any other industrial surface.
Supplier:  Brady™ B30241595OSHADA
 View more versions of this product
Catalog No. 19211119
Only null left
Description
Description
  • Pre-printed headers result in multicolor header signs using a single, black print pass
  • Highly pliable material with high-tack permanent adhesive for rough, highly textured, dirty and harsh environment surfaces—including outdoors
  • Conforms and adheres to "difficult" surfaces like painted cinder blocks, painted wood, textured plastic, paper-jacketed pipes and blow-molded cases
  • Resistant to exposure to chemicals, greases and oils, salt spray, cleaners and detergents and general industrial grime
Specifications
Chemical Identifiers
ASME ANSI Z535, OSHA 1910.145
Vinyl
Pipemarkers, arrow tape, warehouse marking, warning panels, safety signs, arc flash and visual workplace labels
-40°F
-40°C to 82°C
DANGER (Blank label)
BBP30, BBP31, BBP33, BBP35, and BBP37 printers
Permanent Acrylic
180°F
0.004 in.
-40°F to 180 °F
Indoor/Outdoor Tape
Specifications
ASME ANSI Z535, OSHA 1910.145
Vinyl
Permanent Acrylic
2.25 in.
-40°F
3 in.
-40°C to 82°C
5.7 H x 7.6 cm W
Indoor/Outdoor Tape
BBP30, BBP31, BBP33, BBP35, and BBP37 printers
300/Pk.
Pipemarkers, arrow tape, warehouse marking, warning panels, safety signs, arc flash and visual workplace labels
180°F
0.004 in.
76.2 mm
-40°F to 180 °F
DANGER (Blank label)
Red on White
Product Suggestions
Product Suggestions