missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Arc Flash Warning Labels
Description
- Black/orange on white
- W x H: 5 x 3.5 in. (12.7 x 8.89cm)
- Indicate the equipment-specific hazard information
Specifications
Specifications
| Material | Polyester |
| Description | Self-sticking Polyester |
| Mounting Type | Corner Holes |
| Height (English) | 3.5 in. |
| Height (Metric) | 8.89 cm |
| Width (English) | 5 in. |
| Width (Metric) | 12.7 cm |
| Shape | Rectangular |
| Languages | English |
| Dimensions (L x W) | 8.9 x 12.7 cm |
| Show More |