Learn More
Brady™ Anti-Skid Floor Tapes
Catalog No. 19103789
Change view
Click to view available options
Color:
Black
Clear
Yellow
Width (English):
2 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Width (English)
19103789 Clear -
19103790 Black 2 in.
19103779 Yellow -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19103789 Supplier Brady™ Supplier No. 78188
For indoor or outdoor use and floor marking needs
  • Made of rugged B-916 adhesive-backed polyester tape
  • Coated with thousands of particles of durable aluminum oxide grit
  • Bonds permanently to clean, dry floors and withstands grease and oil
Specifications
Color Clear