Learn More
Brady™ Anti-Skid Floor Tapes
For indoor or outdoor use and floor marking needs
Supplier:  Brady™ 78092
 View more versions of this product
Catalog No. 19103779
Only null left
Description
Description
  • Made of rugged B-916 adhesive-backed polyester tape
  • Coated with thousands of particles of durable aluminum oxide grit
  • Bonds permanently to clean, dry floors and withstands grease and oil
Specifications
Specifications
Yellow
Product Suggestions
Product Suggestions