missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Admittance Signs: DANGER - HIGH VOLTAGE

Catalog No. p-169068
Change view
Click to view available options
Material:
Fiberglass
Plastic
Height (English):
7 in.
10 in.
Width (English):
10 in.
14 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Height (English) Width (English)
170462H Fiberglass 7 in. 10 in.
19102031 Plastic 10 in. 14 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 170462H Supplier Brady™ Supplier No. 71565
Only null left

Identifies electrical hazard and high voltage areas

  • Uses bright colors, bold text and intuitive pictograms to warn against entry, machine operation and buried cables
  • Available in three materials - fiberglass, plastic and self-sticking polyester
  • Available in different widths and heights
  • Black/red on white

Specifications

Certifications/Compliance ISO 9001, ANSI Z535, OSHA 1910.145, PSTC-33,
Material Fiberglass
Quantity 1 Ea.
Tensile Strength 9000 psi
Mounting Type Surface Mount
For Use With (Application) Industrial, utility, commercial and institutional environments for both indoor and outdoor use
Height (English) 7 in.
Height (Metric) 17.78 cm
Max. Temperature (English) 193°F
Min. Temperature (English) -40°F
Thickness 0.1 in.
Width (English) 10 in.
Width (Metric) 25.4 cm
Temperature Range (Metric) -40°C to 90°C
Temperature Range (English) -40°F to 193°F
Displayed Text DANGER High Voltage
Product Type Admittance Sign
Color Red on White
Show More Show Less