missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Admittance Sign: DANGER - KEEP OUT HAZARD AREA

Catalog No. 19035909
Change view
Click to view available options
Material:
Plastic
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19035909 Plastic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19035909 Supplier Brady™ Supplier No. 22114

Identify equipment, rooms and materials which contain or are contaminated with hazardous agents

  • Plastic
  • Black/red on white

Specifications

Certifications/Compliance ASTM D 1593, OSHA 1910.145
Material Plastic
Quantity 1 Ea.
Mounting Type Surface Mount
For Use With (Application) Light duty industrial, utility, commercial and institutional environments for both indoor and outdoor use
Height (English) 10 in.
Height (Metric) 25.40 cm
Max. Temperature (English) 175°F
Min. Temperature (English) -40°F
Thickness 0.06 in.
Width (English) 14 in.
Width (Metric) 35.56 cm
Temperature Range (Metric) -40°C to 80°C
Temperature Range (English) -40°F to 175°F
Displayed Text DANGER Keep Out Hazard Area
Product Type Admittance Sign
Color Red on White
Show More Show Less