Learn More
Brady™ Accessories for BMP71 Label Printer
Printer case, Quick charger and Printer battery for use with BMP-71 Label Printer (printer sold separately).
$311.01 - $311.01
Specifications
For Use With (Equipment) BMP71 Label Printer
Sterility Non-sterile
Quantity 1 Ea.
View More Specs
Products 1
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
19-000-211
View Documents
Brady™
M71QC
Quick Charger
Each for $311.01
Only null left
Description
Description
  • BMP™71 Label Printer Accessories from Brady™
  • Printer Case designed to protect your printer against impacts and dust while allowing for easy mobility.
  • Printer Battery offers long battery life - prints up to 3900 1 in. x 2 in. labels per charge, enables cordless printer operation so you can print labels where you need them, when you need them.
  • BMP™71 Quick Charger, charges battery in one hour.
Specifications
Specifications
BMP71 Label Printer
Non-sterile
1 Ea.
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'