missing translation for 'onlineSavingsMsg'
Learn More

Brady™ 65 gal. Spill Kit Refill, Universal

Catalog No. 17111243
Change view
Click to view available options
Capacity (English):
65 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
17111243 65 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17111243 Supplier Brady™ Supplier No. SKA65R

Universal Spill Kit Refill is intended for use with the Allwik™ 65 gal. universal spill kit. (Refill kit only—container sold separately.)

  • Highly versatile, grey absorbents pick up water-based, petroleum-based and non-aggressive, chemical fluids
  • Improve worker productivity and efficiency by placing spill kits around your facility close to areas prone to spills
  • Spill kits are ideal for fast response to critical accidents

Specifications

Color Gray
Capacity (English) 65 gal.
Capacity (Metric) 246 L
Type Universal Spill Kit Refill
For Use With (Application) Universal - All purpose spills
Includes (100) 15 x 19 in. pads, (7) 3 in. x 12 ft. socks, (7) 18 x 18 in. pillows, 1 pair goggles, 1 pair Nitrile gloves, 3 disposal bags, handbook