missing translation for 'onlineSavingsMsg'
Learn More

Brady™ 65 gal. Spill Kit Refill, Oil Only

Catalog No. 17111242
Change view
Click to view available options
Capacity (English):
65 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
17111242 65 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17111242 Supplier Brady™ Supplier No. SKO65R

Oil-Only Spill Kit Refill is intended for use with the Allwik™ 65 gal. spill kit for oil-based materials. (Refill kit only—container sold separately.)

  • White absorbents pick up petroleum-based fluids, paints and non-water soluble chemicals while repelling water - great for indoor and outdoor use
  • Improve worker productivity and efficiency by placing spill kits around your facility close to areas prone to spills
  • Spill kits are ideal for fast response to critical accidents

Specifications

Color White
Capacity (English) 65 gal.
Capacity (Metric) 246 L
Type Spill Kit Refill, Oil-only
For Use With (Application) Oil spills
Includes (100) 15 x 19 in. pads, (7) 3 in. x 12 ft. socks, (7) 18 x 18 in. pillows, 1 pair goggles, 1 pair Nitrile gloves, 3 disposal bags, handbook