missing translation for 'onlineSavingsMsg'
Learn More

Poltex Bracket for Pipette Controller

The Poltex Bracket for Pipette Controller is ideal for hanging pipette controller securely up and out of the way.

Supplier:  Poltex BRKM

Encompass

 View more versions of this product

Catalog No. 22000242


Only null left

Description

Description

  • Pipette Controller hole is 1.125 in. diameter
  • PETG construction
  • Available mounting options include VHB tape, powerful magnets, or keyhole slots for wall mount
Specifications

Chemical Identifiers

63.5 mm
114 mm
70 mm

Specifications

Magnet
63.5 mm
114 mm
70 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions