missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Bovine Rhodopsin (aa 338-348) Synthetic Peptide
Description
Bovine Rhodopsin (residues 338-348) Synthetic Peptide, PEP-174, from Invitrogen.
Visual pigments such as rhodopsin and porphyropsin are light-absorbing molecules that mediate vision. Rhodopsin consists of an apoprotein, opsin, covalently linked to 11-cis-retinal. This receptor is coupled to the activation of phospholipase C. Porphyropsin consists of opsin covalently linked to 11-cis 3,4-didehydroretinal.
Specifications
Specifications
| Gene ID (Entrez) | 509933 |
| Species | Bovine |
| Purification Method | purified |
| Accession Number | P02699 |
| Common Name | Bovine Rhodopsin (aa 338-348) |
| Concentration | 0.5 mg/mL |
| Content And Storage | -20°C. Avoid freeze-thaw cycles. |
| Description | Synthetic Peptide |
| Format | Lyophilized |
| Formulation | peptide with no preservative |
| Show More |