missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Bovine Pituitary Extract

Catalog No. MP092850450
Change view
Click to view available options
Quantity:
2.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP092850450 2.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP092850450 Supplier MP Biomedicals™ Supplier No. 092850450
Only null left

Filtered Bovine Pituitary Extract for cell culture.

  • Bovine Pituitary Extract (BPE) is made from fresh, flash-frozen pituitary glands from young, healthy cattle between the ages of 18 to 22 months
  • The cattle are born, raised and slaughtered in New Zealand from a USDA approved source
  • New Zealand cattle are internationally recognized as free from bovine spongiform encephalopathy (BSE, mad-cow disease) and have no history of blue tongue disease
  • BPE continues to be the most successful of the tissue-sourced extracts to date and allows researchers to study normal primary cell types including stem cells in proliferating cultures
  • Formulated at 14 mg/mL crude protein, this aqueous BPE has a full spectrum of mitogens and differentiation factors
  • The Bovine Pituitary Extract is not lyophilized, as this process has been shown to reduce biological activity
  • For cell culture, the optimal medium supplementation rate is 35 μg/mL BPE based on growth tests on human mammary epithelial cells, epidermal keratinocytes, prostatic and bronchial epithelial cells
  • This product has been sterile-filtered prior to shipping

Specifications

Content And Storage Store at -20°C, do not thaw and refreeze
Concentration 14 mg/mL
Form Liquid
Sterility Sterile
For Use With (Application) Cell Culture
Quantity 2.5 mL

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.