missing translation for 'onlineSavingsMsg'
Learn More

Bouins Fluid, Fisherbrand™

Catalog No. 11002230
Change view
Click to view available options
Quantity:
1 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11002230 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 11002230 Supplier Statlab Supplier No. 11002230

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Harmful if swallowed, in contact with skin, or if inhaled

A good lymph node and glycogen fixative as well as a superior trichome stain mordant.

  • With Bouins Fluid, soft and delicate structures are preserved and trichrome stains will be brilliant with color while demonstrating crisp nuclear detail.
  • Bouin's Fluid is also an excellent fixative to use on biopsy specimens of the gastrointestinal tract because the nuclear staining is superior to 10% formalin.

Specifications

For Use With (Equipment) All Tissue Processors
Grade Histological
Product Type Fixative
Quantity 1 gal.
Solution Type Fixative
Form Solution

Made in the USA