missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bouin's Fixative Solution, Picric Acid-Formalin-Acetic Acid Mixture, Ricca Chemical
$119.03 - $2,605.50
Specifications
| Chemical Name or Material | Bouin's Fixative Solution |
|---|---|
| Physical Form | Liquid |
| Name Note | Picric Acid-Formalin-Acetic Acid Mixture |
| CAS | 7732-18-5,50-00-0,64-19-7,67-56-1,88-89-1 |
| CAS Min % | 68.0,24.33,4.76,0.87 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
112016
|
Ricca Chemical Company
112016 |
500 mL | Natural Poly Bottle |
N/A
|
|
||||
|
11205
|
Ricca Chemical Company
11205 |
20 L | Cubitainer |
N/A
|
|
||||
Specifications
| Bouin's Fixative Solution | |
| Picric Acid-Formalin-Acetic Acid Mixture | |
| 68.0,24.33,4.76,0.87 | |
| Yellow |
| Liquid | |
| 7732-18-5,50-00-0,64-19-7,67-56-1,88-89-1 | |
| 71.0,25.33,4.96,0.91 | |
| Laboratory |