missing translation for 'onlineSavingsMsg'
Learn More

VAI Lightweight Bouffant Hats

Bouffant Hats are designed and manufactured in a pleated design that is low in lot and full coverage, for use in a cleanroom setting.

$258.50 - $258.50

Specifications

Product Type Cleanroom Bouffant
Material Polypropylene
Color Blue
Disposable Single-use
Latex Free Yes
View More Specs

Products 1
Catalog Number Mfr. No. Size Diameter (Metric) Diameter (English) Price Quantity  
Catalog Number Mfr. No. Size Diameter (Metric) Diameter (English) Price Quantity  
19547268
View Documents
VAI
150BFNS1530
24 in. 61 cm 24 in.
Case of 1000 Each for $258.50
Only null left
 
Description

Description

  • Individually inspected for size, stitching, and workmanship
  • Neatly stacked and cleanroom packaged
  • Low linting fabric
Specifications

Specifications

Cleanroom Bouffant
Blue
Yes
100 Pk., 10 Pk./Cs.
Polypropylene
Single-use
Non-sterile
Class 1000 (ISO 6)
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'