missing translation for 'onlineSavingsMsg'
Learn More

International Enviroguard™ Flat Bouffant Caps

Catalog No. p-7222262
Change view
Click to view available options
Color:
White
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19472790 White
19472789 White
19472791 -
19472792 White
19472793 -
19472794 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19472790 Supplier International Enviroguard™ Supplier No. S110NW19
Only null left

Flat bouffant caps provide protection against dirt, grime, and particulates in non-hazardous environments. Available in blue or white in a variety of sizes.

  • Provide a particulate barrier and helps prevent hair from contaminating a product or work environment
  • Low-linting, lightweight cover restrains hair and offers great ventilation
  • Designed to protect against dirt, grime, and particulates in non-hazardous environments
  • Flat style
  • Latex-free elastic
  • Possible applications include food processing, construction, pharmaceuticals, environmental cleanup, and more

Specifications

Product Type Bouffant
Material Polypropylene
Color White
Disposable Single-use
Model S110NW
Diameter (English) 19 in.
For Use With (Application) Food Processing, Construction, Pharmaceuticals, Environmental Cleanup