missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wanzl Bottom Shelf with Black Powdercoat Finish
Description
- Bottm shelf
- Replacement part for bulk shelving system
- Black powdercoat finish
Specifications
Specifications
| Width (English) | 48 in. |
| For Use With (Application) | Shelving |
| Shelf Type | Bottom Shelf |
| No. of Shelves | 1 |
| Finish | Powdercoat |
| Dimensions (L x W) | 36 x 48 in. |
| Color | Black |
| Material | Steel |