missing translation for 'onlineSavingsMsg'
Learn More

Wanzl Bottom Shelf with Black Powdercoat Finish

Catalog No. 11222152
Change view
Click to view available options
Dimensions (L x W):
24 x 36 in.
24 x 48 in.
36 x 36 in.
36 x 48 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
11222152 24 x 36 in.
11222153 24 x 48 in.
11222154 36 x 36 in.
11222155 36 x 48 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 11222152 Supplier Wanzl Supplier No. T.BCRSHB2436B
Only null left

Bottom Shelf for bulk shelving.

  • Bottm shelf
  • Replacement part for bulk shelving system
  • Black powdercoat finish

Specifications

Width (English) 36 in.
For Use With (Application) Shelving
Color Black
Finish Powdercoat
No. of Shelves 1
Shelf Type Bottom Shelf
Dimensions (L x W) 24 x 36 in.
Material Steel