missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Bottle Top Dispenser with Re-Circulation Valve

Provides a contemporary design coupled with a recirculation valve. Bottle Top Dispenser with Re-Circulation Valve has enhanced chemical compatibility and smooth piston movement.

Specifications

Product Type Bottletop Dispenser
Autoclavable Autoclavable
Color White and Fisher Blue
Density 2.2 g/cm3
Material Polypropylene - Outer Body (10% Glass Filled)
View More Specs

Products 1
Catalog Number Volume (Metric) Price Quantity & Availability  
Catalog Number Volume (Metric) Price Quantity & Availability  
13681527
View Documents Promotion Details
2.5 to 30 mL N/A
N/A
Description

Description

  • Re-circulation valve helps user in quick purging without wasting any reagent
  • Specially designed unique spring-less valve provides greater chemical compatibility, no chemical deposition and minimizing the maintainence
  • Presence of unique spring-less valve, PTFE piston and ETP O-ring provides greater chemical compatibility, ensures wiper like smooth and effortless piston movement
  • Specially designed 180° rotation volume setting knob provides quick and effortless volume adjustment
  • Adjustable delivery nozzle provides flexibility to users in demanding laboratory conditions
  • Bottle neck adapters of different sizes (28, 30, 32, 38, 40 and 45mm) ensures compatibility of dispenser with majority of reagent bottles
  • Each dispenser is accompanied with tool, which helps in easy and quick re-calibration
  • Contemporary and well designed contours, silky surface finish
  • Individually calibrated as per ISO 8655-2
  • Autoclavable 
Storage:
  • Store instrument and accessories only in clean conditions in cool and dry place (from -20°C to 50°C)
Specifications

Specifications

Bottletop Dispenser
White and Fisher Blue
Polypropylene - Outer Body (10% Glass Filled)
Calibration tool, Adapters set (5 nos.), Instruction Manual, Inlet tube, Calibration report
500mm2
Autoclavable
2.2 g/cm3
Bottle Top Dispenser is used in laboratories to dispense reagent from the bottle in a preset volume accurately and safely into a receiving vessel.
300mbar to 600mbar
1 year free replacement warranty against any manufacturing defects.
Videos
SDS
Documents

Documents