missing translation for 'onlineSavingsMsg'
Learn More

BRANDTECH™ BRAND™ Bottle Stand for BRAND™ Bottletop Dispensers and Burettes

This bottle Stand from BRAND™ is designed to assist in maintaining a secure setup during bottletop dispensing.

Supplier:  BRANDTECH™ BRAND™ 704275

Catalog No. 13880713


Only null left

Description

Description

  • Polypropylene bottle stand provides chemical resistance and stability while using Dispensette™, Titrette™, and other bottletop dispensers
  • Includes 325 mm support rod
  • Baseplate is 220 mm x 160 mm
  • Weight is 1.13 kg.
Specifications

Specifications

Bottletop Stand
A variety of bottle sizes
Polypropylene
225 x 160 x 325 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions