Learn More
Bottle Rocket Launcher
Catalog No. S41988
Change view
Click to view available options
Product Type:
Bottle Rocket Launcher
Catalog No. Product Type
S41988 Bottle Rocket Launcher
1 options
Catalog No. S41988 Supplier Customer Channels Group (Ccg) Supplier No. 17000
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes
Instructions and a paper astrolabe for determining rocket altitude
Get a quantitative idea of the effects of varying moments of inertia.
Pressurize a plastic soda bottle with an ordinary bicycle pump (not included). Vary the amount of pressure and/or water fuel to see whose rocket will go the highest. Experiment with different fin styles to observe how changes in design affect flight paths.
  • Solid aluminum construction
  • Simple triggering mechanism
  • Mounting stakes
Specifications
Product Type Bottle Rocket Launcher
For Use With (Application) Explores the basic principles of rocketry and Newton's third law
Includes Instructions and a paper astrolabe for determining rocket altitude