Learn More
Chemglass Inc 950mL Bottle, 53-400 GPI Thread, Amber Glass
Catalog No. CG82005
Encompass
950mL Bottle, 53-400 GPI Thread, Amber Glass. These round, wide mouth amber glass bottles are used for storage of light sensitive chemicals. The 50mL and 120mL bottles have a black phenolic cap with a polyethylene cone shaped liner. Has a white polypropylene cap with a polyethylene faced foam liner. Sold with cap attached to the bottle in case quantities only.
Catalog No. CG82005 Supplier Chemglass Inc Supplier No. CG82005
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
950mL Bottle, 53-400 GPI Thread, Amber Glass. These round, wide mouth amber glass bottles are used for storage of light sensitive chemicals. The 50mL and 120mL bottles have a black phenolic cap with a polyethylene cone shaped liner. Has a white polypropylene cap with a polyethylene faced foam liner. Sold with cap attached to the bottle in case quantities only.