Learn More
BRANDTECH™ BRAND™ Bottle Adapters for Dispensette™ and seripettor™ Bottletop Dispensers and Titrette™ Bottletop Burettes
Adapters for common bottle threads and standard taper joints
Supplier:  BRANDTECH™ BRAND™ 704340
 View more versions of this product
Catalog No. 13880746
Only null left
Description
Description
  • Economical polypropylene adapters work for most applications
  • ETFE/PTFE adapters for aggressive acids and solvents
  • Leading number refers to male thread of adapter
  • Second number refers to bottle neck the adapter fits
  • 28 mm, 24 mm and 22 mm bottle threads require stacking adapter with 45/33 mm adapter.
Specifications
Specifications
Polypropylene
External Thread: 33 mm; Bottle Thread: 40 mm
33 mm
ottle Thread: 40 mm
40 mm
Product Suggestions
Product Suggestions