Learn More
BRANDTECH™ BRAND™ Bottle Adapters for Dispensette™ and seripettor™ Bottletop Dispensers and Titrette™ Bottletop Burettes
Adapters for common bottle threads and standard taper joints
Supplier: BRANDTECH™ BRAND™ 704375
Description
- Economical polypropylene adapters work for most applications
- ETFE/PTFE adapters for aggressive acids and solvents
- Leading number refers to male thread of adapter
- Second number refers to bottle neck the adapter fits
- 28 mm, 24 mm and 22 mm bottle threads require stacking adapter with 45/33 mm adapter.
Specifications
| PTFE, ETFE | |
| External Thread: 33mm; Bottle Thread: 24 mm | |
| 33 mm |
| Bottle Thread: 24 mm | |
| 24 mm |