Learn More
Bosutinib, Thermo Scientific™
Chemical Identifiers
| CAS | 380843-75-4 |
|---|---|
| Molecular Formula | C26H29Cl2N5O3 |
| Molecular Weight (g/mol) | 530.45 |
| MDL Number | MFCD07367846 |
| InChI Key | UBPYILGKFZZVDX-UHFFFAOYSA-N |
| IUPAC Name | 4-[(2,4-dichloro-5-methoxyphenyl)amino]-6-methoxy-7-[3-(4-methylpiperazin-1-yl)propoxy]quinoline-3-carbonitrile |
| SMILES | COC1=CC(NC2=C3C=C(OC)C(OCCCN4CCN(C)CC4)=CC3=NC=C2C#N)=C(Cl)C=C1Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC465650010
|
Thermo Scientific Chemicals
465650010 |
1 g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC465652500
|
Thermo Scientific Chemicals
465652500 |
250 mg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 380843-75-4 | |
| 530.45 | |
| UBPYILGKFZZVDX-UHFFFAOYSA-N | |
| COC1=CC(NC2=C3C=C(OC)C(OCCCN4CCN(C)CC4)=CC3=NC=C2C#N)=C(Cl)C=C1Cl |
| C26H29Cl2N5O3 | |
| MFCD07367846 | |
| 4-[(2,4-dichloro-5-methoxyphenyl)amino]-6-methoxy-7-[3-(4-methylpiperazin-1-yl)propoxy]quinoline-3-carbonitrile |
Specifications
| 380843-75-4 | |
| MFCD07367846 | |
| Solubility aq. soln.: .. Other solubilities: soluble in methanol and dmso | |
| COC1=CC(NC2=C3C=C(OC)C(OCCCN4CCN(C)CC4)=CC3=NC=C2C#N)=C(Cl)C=C1Cl | |
| 530.45 | |
| Bosutinib |
| C26H29Cl2N5O3 | |
| 1 g | |
| UBPYILGKFZZVDX-UHFFFAOYSA-N | |
| 4-[(2,4-dichloro-5-methoxyphenyl)amino]-6-methoxy-7-[3-(4-methylpiperazin-1-yl)propoxy]quinoline-3-carbonitrile | |
| 530.45 |
Safety and Handling
GHS H Statement May cause an allergic skin reaction.
Causes serious eye irritation.
GHS P Statement IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Warning
RUO â Research Use Only