missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synthware™ Bosshead
Bosshead for attaching clamps to hood rack system
Supplier: Synthware™ J111200
Description
Designed for use with rods ranging from 15 to 19mm in diameter.
Spécifications
| Knobless | |
| Chrome-plated Metal | |
| Organic Synthesis | |
| Fits 19 mm Rod |
| Bosshead Clamp | |
| Bosshead only | |
| No knobs | |
| 19 mm |