Learn More
Eisco™ Boss Head Clamp, Die Cast Alloy, Powder Coated Finish, up to 16mm Rods
Catalog No. s24263
Change view
Click to view available options
Color:
Gray
Catalog No. Color
S24263 Gray
1 options
Catalog No. S24263 Supplier Eisco™ Supplier No. CH0663A
Only null left
For use with laboratory stands, this popular boss head is made of die casted alloy with a powder coat finish. It accepts rods up to 16mm in diameter. Metal thumbscrews allow for adjustment. Measures approx 11x3x3cm.
Specifications
Type Clamp
Material Metal
Color Gray
Jaw Type Fixed