missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Borosilicate Glass Superloops

Catalog No. 361041166
Change view
Click to view available options
Volume (Metric):
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
361041166 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 361041166 Supplier Cytiva Supplier No. 18111382

Use for the introduction of large sample volumes into a pressurised fluid LC system. Cytiva Lifescience™ Borosilicate Glass Superloops work in conjunction with 7-port valves.

  • Superloop can be used in conjunction with 7-port valves. The patented devices enable controlled loading of large samples, either in the form of multiple small injections or a single large injection. They may also be used to inject pulses of a specific buffer solution.
  • Flexible: inject large or small sample volumes.
  • High reproducibility and recovery.
  • No gradient dilution with internal movable seal.

Specifications

Volume (Metric) 50 mL
Packaging Quantity 1 Each
Pressure 580 psi