missing translation for 'onlineSavingsMsg'
Learn More

Boron trifluoride-tetrahydrofuran complex, BF{3} 45.5%

Catalog No. AAL1759322
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1759322 100 g
AAL1759336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1759322 Supplier Thermo Scientific Chemicals Supplier No. L1759322
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 462-34-0 | C4H8BF3O | 139.91 g/mol

Boron trifluoride-tetrahydrofuran complex is used as a polymerization initiator and as a dopant in the semiconductor industry. It serves as a reagent for deoxofluorination of alcohols and carbonyls. Further, it is used in the synthesis of methacrylate-PEG-methacrylate-boron trifluoride self-doped gel polymer electrolytes and hydroboration-oxidation of divinylferrocene. In addition to this, it is used as a reagent for the preparation of NK1 receptor antagonists, complex polyketide natural products and selective serotonin reuptake inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Boron trifluoride-tetrahydrofuran complex is used as a polymerization initiator and as a dopant in the semiconductor industry. It serves as a reagent for deoxofluorination of alcohols and carbonyls. Further, it is used in the synthesis of methacrylate-PEG-methacrylate-boron trifluoride self-doped gel polymer electrolytes and hydroboration-oxidation of divinylferrocene. In addition to this, it is used as a reagent for the preparation of NK1 receptor antagonists, complex polyketide natural products and selective serotonin reuptake inhibitors.

Solubility
Miscible with dimethyl formamide.

Notes
Incompatible with strong oxidizing agents and water.

Chemical Identifiers

CAS 462-34-0
Molecular Formula C4H8BF3O
Molecular Weight (g/mol) 139.91
MDL Number MFCD00040372
InChI Key CHNLPLHJUPMEOI-UHFFFAOYSA-N
Synonym boron trifluoride tetrahydrofuran complex, trifluoro tetrahydro-1h-furan-1-ium-1-yl borate, pubchem7062, trifluoro oxolan-1-ium-1-yl boranuide, borontrifluoride tetrahydrofuran complex, boron trifluoride tetrahydrofuran complex, contains sulfur dioxide as peroxide formation suppressor
PubChem CID 60146129
SMILES FB(F)F.C1CCOC1

Specifications

Melting Point 8°C to 9°C
Density 1.268
Boiling Point 180°C
Flash Point 92°C (197°F)
Quantity 100 g
UN Number UN2922
Sensitivity Moisture sensitive
Solubility Information Miscible with dimethyl formamide.
IUPAC Name oxolane; trifluoroborane
Formula Weight 139.91
Assay BF3 45.5%
Chemical Name or Material Boron trifluoride-tetrahydrofuran complex
Show More Show Less
Hazard Category H227-H314-H332-H372
Hazard Statement GHS H Statement
H301-H330-H372-H314-H318-H227
Toxic if swallowed.
Fatal if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P270-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 207-325-9
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only