missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Boron trifluoride-methanol solution

Catalog No. B1252250ML
Encompass
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1252250ML 250 mL
B12521L 1 L
B1252500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B1252250ML Supplier Sigma Aldrich Supplier No. B1252250ML
Only null left

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Boron trifluoride-methanol (BF-M) is an esterification reagent. BF-M is a powerful acidic catalyst for the esterification of the fatty acids. It easily cleaves acetylated dyes resulting in free amino groups. BF-M catalyzed trans esterification of vegetable oils forms biodiesel.Used in the deactylation of acetanilides.It is also used to study the synthesis of amino group containing cyanine dyes of different types and to prepare fatty acid methyl esters.

Specifications

CAS 373-57-9
Density 0.859 g/mL (at 25°C)
Molecular Formula CH4BF3O
Linear Formula BF3 -+ MeOH
Quantity 250 mL
MDL Number MFCD00071635
Synonym Trifluoroborane
Molecular Weight (g/mol) 99.85
Percent Purity 12-16% (NaOH, titration)