missing translation for 'onlineSavingsMsg'
Learn More

Boron trifluoride-Methanol Solution, ∼10 (∼1.3 M), MilliporeSigma™ Supelco™

Catalog No. p-7187944
Change view
Click to view available options
Quantity:
1 mL
10 mL
100 mL
10 x 1 mL
250 mL
1 L
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020687 1 mL
111020140 10 x 1 mL
111020608 10 mL
111020334 100 mL
111019291 250 mL
111018152 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 111020687 Supplier Merck Emd Millipore Supplier No. 157161ML
Only null left

Chemical Identifiers

MDL Number MFCD00071635

Specifications

CAS 373-57-9
Quantity 1 mL
Linear Formula BF3 · MeOH
Flash Point 9°C
Formula Weight 99.85
Grade GC Derivatization
Concentration or Composition (by Analyte or Components) 10% (1.3 M)
Density 0.86 g/mL (at 20°C)