missing translation for 'onlineSavingsMsg'
Learn More

Boron Trifluoride - Ethyl Ether Complex 98.0+%, TCI America™

Catalog No. b0527500ml
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
25 mL
500 mL
Catalog No. Quantity
B0527500ML 500 mL
B052725ML 25 mL
B0527100ML 100 mL
3 options

Catalog No. B0527500ML

Supplier: TCI America B0527500ML

Only null left

Chemical Identifiers

CAS 109-63-7
Molecular Formula C4H10BF3O
Molecular Weight (g/mol) 141.93
MDL Number MFCD00013194
InChI Key KZMGYPLQYOPHEL-UHFFFAOYSA-N
PubChem CID 8000
IUPAC Name ethoxyethane; trifluoroborane
SMILES FB(F)F.CCOCC

Specifications

Melting Point -60°C
Color Yellow
Boiling Point 124°C
Quantity 500 mL
UN Number 2604
Formula Weight 141.93
Percent Purity ≥98.0% (W)
Physical Form Liquid
Chemical Name or Material Boron Trifluoride - Ethyl Ether Complex
EINECSNumber (2)-2107
RTECSNumber ED2275000
TSCA Yes