missing translation for 'onlineSavingsMsg'
Learn More

Boron Trifluoride - Butyl Ether Complex (BF3 ca. 30%), TCI America™

Catalog No. B207425ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B207425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B207425ML Supplier TCI America Supplier No. B207425ML

Chemical Identifiers

CAS 593-04-4
Molecular Formula C8H18BF3O
Molecular Weight (g/mol) 198.04
MDL Number MFCD00013195
InChI Key PVKVBDZRILNPJY-UHFFFAOYSA-N
Synonym 1-butoxybutane; trifluoroborane, boron trifluoride dibutyl etherate, boron trifluoride-butyl ether complex, boron, trifluoro 1,1'-oxybis butane-, t-4, boron trifluoride; di-n-butyl ether, 1-butoxybutane-trifluoroborane 1:1, 1-butoxybutane; tris fluoranyl borane, trifluoro 1,1'-oxybis butane boron
PubChem CID 68970
IUPAC Name 1-butoxybutane; trifluoroborane
SMILES FB(F)F.CCCCOCCCC

Specifications

Quantity 25 mL
UN Number 3265
Formula Weight 198.04
Physical Form Liquid
Chemical Name or Material Boron Trifluoride - Butyl Ether Complex (BF3 ca. 30%)
EINECSNumber (2)-2107
RTECSNumber ED2275000
TSCA Yes