missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boron trifluoride - acetic acid complex, ca. 33% BF3, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 277220250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Lewis acidSpecifications
| Boron trifluoride - acetic acid complex | |
| 373-61-5 | |
| 140°C to 148°C | |
| Glass bottle | |
| BF3·2CH3COOH | |
| 25mL | |
| 16,176 | |
| boron trifluoride acetic acid complex, bf3.c4h8o4, acetic acid-boron trifluoride complex, acetic acid boron trifluoride 2:1, bis acetic acid ; boron trifluoride, boron trifluoride acetic acid complex, technical bf3 basis, borate 1-, bis acetato-o difluoro-, dihydrogen fluoride, beta-4, borate 1-, bis acetato-.kappa.o difluoro-, hydrogen fluoride 1:2:1 , t-4 | |
| COTMJGCQSLZICX-UHFFFAOYSA-N | |
| acetic acid;trifluoroborane | |
| 11116786 | |
| ca. 33% BF3 |
| ca. 33% BF3 | |
| 1.3450g/mL | |
| 84°C | |
| BF3·C4H8O4 | |
| MFCD00036359 | |
| 04, 115 | |
| 1.345 | |
| Solubility in water: reacts | |
| B(F)(F)F.CC(=O)O.CC(=O)O | |
| 187.92 | |
| 187.92 |
RUO â Research Use Only