Learn More
Boron trifluoride - acetic acid complex, ca. 33% BF3
CAS: 373-61-5 | C4H8BF3O4 | 187.91 g/mol
$719.20 - $719.20
Chemical Identifiers
| CAS | 373-61-5 |
|---|---|
| Molecular Formula | C4H8BF3O4 |
| Molecular Weight (g/mol) | 187.91 |
| MDL Number | MFCD00036359 |
| InChI Key | OVYKFNOIPQYMRU-UHFFFAOYSA-K |
| Synonym | boron trifluoride acetic acid complex, bf3.c4h8o4, acetic acid-boron trifluoride complex, acetic acid boron trifluoride 2:1, bis acetic acid ; boron trifluoride, boron trifluoride acetic acid complex, technical bf3 basis, borate 1-, bis acetato-o difluoro-, dihydrogen fluoride, beta-4, borate 1-, bis acetato-.kappa.o difluoro-, hydrogen fluoride 1:2:1 , t-4 |
| PubChem CID | 11116786 |
| IUPAC Name | acetic acid;trifluoroborane |
| SMILES | [H+].[H+].[B+3].[F-].[F-].[F-].CC([O-])=O.CC([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC277225000
|
Thermo Scientific Chemicals
277225000 |
500 mL | Glass bottle |
Each for $719.20
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acidChemical Identifiers
| 373-61-5 | |
| 187.91 | |
| OVYKFNOIPQYMRU-UHFFFAOYSA-K | |
| 11116786 | |
| [H+].[H+].[B+3].[F-].[F-].[F-].CC([O-])=O.CC([O-])=O |
| C4H8BF3O4 | |
| MFCD00036359 | |
| boron trifluoride acetic acid complex, bf3.c4h8o4, acetic acid-boron trifluoride complex, acetic acid boron trifluoride 2:1, bis acetic acid ; boron trifluoride, boron trifluoride acetic acid complex, technical bf3 basis, borate 1-, bis acetato-o difluoro-, dihydrogen fluoride, beta-4, borate 1-, bis acetato-.kappa.o difluoro-, hydrogen fluoride 1:2:1 , t-4 | |
| acetic acid;trifluoroborane |
Specifications
| 373-61-5 | |
| Brown-Yellow to Yellow | |
| 140°C to 148°C | |
| Glass bottle | |
| BF3·2CH3COOH | |
| 500 mL | |
| 16,176 | |
| boron trifluoride acetic acid complex, bf3.c4h8o4, acetic acid-boron trifluoride complex, acetic acid boron trifluoride 2:1, bis acetic acid ; boron trifluoride, boron trifluoride acetic acid complex, technical bf3 basis, borate 1-, bis acetato-o difluoro-, dihydrogen fluoride, beta-4, borate 1-, bis acetato-.kappa.o difluoro-, hydrogen fluoride 1:2:1 , t-4 | |
| OVYKFNOIPQYMRU-UHFFFAOYSA-K | |
| acetic acid;trifluoroborane | |
| 11116786 | |
| ca. 33% BF3 | |
| Boron trifluoride - acetic acid complex, ca. 33% BF3 |
| -47°C | |
| 1.3450g/mL | |
| 84°C | |
| C4H8BF3O4 | |
| MFCD00036359 | |
| 04, 115 | |
| 1.345 | |
| Solubility in water: reacts | |
| [H+].[H+].[B+3].[F-].[F-].[F-].CC([O-])=O.CC([O-])=O | |
| 187.91 | |
| 187.92 | |
| Liquid |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 206-768-5
TSCA : TSCA
Recommended Storage : Product darkens on standing
RUO – Research Use Only