missing translation for 'onlineSavingsMsg'
Learn More

Boron trifluoride - acetic acid complex, ca. 33% BF3

Catalog No. AC277225000
Change view
Click to view available options
Quantity:
25 mL
500 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC277225000 500 mL Glass bottle
AC277220250 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC277225000 Supplier Thermo Scientific Chemicals Supplier No. 277225000

CAS: 373-61-5 | C4H8BF3O4 | 187.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid

Chemical Identifiers

CAS 373-61-5
Molecular Formula C4H8BF3O4
Molecular Weight (g/mol) 187.91
MDL Number MFCD00036359
InChI Key OVYKFNOIPQYMRU-UHFFFAOYSA-K
Synonym boron trifluoride acetic acid complex, bf3.c4h8o4, acetic acid-boron trifluoride complex, acetic acid boron trifluoride 2:1, bis acetic acid ; boron trifluoride, boron trifluoride acetic acid complex, technical bf3 basis, borate 1-, bis acetato-o difluoro-, dihydrogen fluoride, beta-4, borate 1-, bis acetato-.kappa.o difluoro-, hydrogen fluoride 1:2:1 , t-4
PubChem CID 11116786
IUPAC Name acetic acid;trifluoroborane
SMILES [H+].[H+].[B+3].[F-].[F-].[F-].CC([O-])=O.CC([O-])=O

Specifications

Melting Point -47°C
Color Brown-Yellow to Yellow
Density 1.3450g/mL
Boiling Point 140°C to 148°C
Flash Point 84°C
Packaging Glass bottle
Linear Formula BF3·2CH3COOH
Quantity 500 mL
Beilstein 04, 115
Fieser 16,176
Specific Gravity 1.345
Solubility Information Solubility in water: reacts
Formula Weight 187.92
Percent Purity ca. 33% BF3
Physical Form Liquid
Chemical Name or Material Boron trifluoride - acetic acid complex, ca. 33% BF3
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 206-768-5
TSCA TSCA
Recommended Storage Product darkens on standing

RUO – Research Use Only