missing translation for 'onlineSavingsMsg'
Learn More

Boron trifluoride-acetic acid complex,35-36.5% BF3

Catalog No. p-7081474
Change view
Click to view available options
Quantity:
250 mL
1000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA11471AK 250 mL
AAA11471AU 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA11471AK Supplier Thermo Scientific Chemicals Supplier No. A11471AK
Only null left

CAS: 373-61-5 | C4H8BF3O4 | 187.91 g/mol

It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff. It is also used as a desilylating agent. It is also been used in the protodesilylation of the phenyl group as part of the conversion of the dimethylphenylsilyl group into a hydroxyl group with retention of configuration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff. It is also used as a desilylating agent. It is also been used in the protodesilylation of the phenyl group as part of the conversion of the dimethylphenylsilyl group into a hydroxyl group with retention of configuration.

Solubility
It reacts with water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Store under inert gas. Stable, but moisture sensitive. Incompatible with water, moist air, metals, strong oxidizing agents. Store under dry atmosphere.

Chemical Identifiers

CAS 373-61-5
Molecular Formula C4H8BF3O4
Molecular Weight (g/mol) 187.91
MDL Number MFCD00036359
InChI Key OVYKFNOIPQYMRU-UHFFFAOYSA-K
Synonym boron trifluoride acetic acid complex, bf3.c4h8o4, acetic acid-boron trifluoride complex, acetic acid boron trifluoride 2:1, bis acetic acid ; boron trifluoride, boron trifluoride acetic acid complex, technical bf3 basis, borate 1-, bis acetato-o difluoro-, dihydrogen fluoride, beta-4, borate 1-, bis acetato-.kappa.o difluoro-, hydrogen fluoride 1:2:1 , t-4
PubChem CID 11116786
IUPAC Name acetic acid;trifluoroborane
SMILES [H+].[H+].[B+3].[F-].[F-].[F-].CC([O-])=O.CC([O-])=O

Specifications

Melting Point -47°C
Density 1.353
pH <1
Boiling Point 143°C to 146°C
Flash Point 83°C (181°F)
Odor Pungent
Refractive Index 1.367
Quantity 250 mL
UN Number UN1742
Beilstein 3686177
Sensitivity Moisture sensitive
Solubility Information It reacts with water.
Formula Weight 187.91
Percent Purity 96%
Chemical Name or Material Boron trifluoride-acetic acid complex
Show More Show Less
Hazard Category H227-H302-H314
Hazard Statement GHS H Statement
H301-H330-H314-H318-H227
Toxic if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: BORON TRIFLUORIDE ACETIC ACID COMPLEX, LIQUID
EINECSNumber 206-768-5
RTECSNumber ED2280000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only