missing translation for 'onlineSavingsMsg'
Learn More

Boron trifluoride, 12% (1.5M) in methanol, AcroSeal™

Catalog No. AC445271000
Change view
Click to view available options
Quantity:
100 mL
800 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC445271000 100 mL
AC445278000 800 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC445271000 Supplier Thermo Scientific Chemicals Supplier No. 445271000

CAS: 373-57-9 | BF3 | 67.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid

Specifications

Boiling Point 65.0°C
Chemical Name or Material Boron trifluoride
Melting Point -98.0°C
CAS 373-57-9, 67-56-1
Quantity 100 mL
Molecular Formula BF3
Linear Formula BF3
MDL Number MFCD00011316
Flash Point 4°C
Synonym boron trifluoride-methanol solution, boron trifluoride-methanol complex, boron trifluoride methanol, boron trifluoride-methanol solution in methanol, bf3 methanol, .bf3 in methanol, boron fluoride methanol, bf3 meoh, borontrifluoride methanol, boron trifluoride solution
Solubility Information Solubility in water: may decompose
InChI Key WTEOIRVLGSZEPR-UHFFFAOYSA-N
SMILES FB(F)F
IUPAC Name trifluoroborane
Molecular Weight (g/mol) 67.81
PubChem CID 11062313
Formula Weight 67.81
Packaging AcroSeal™ Glass bottle
Specific Gravity 0.87
Density 0.8700g/mL
Show More Show Less

RUO – Research Use Only