missing translation for 'onlineSavingsMsg'
Learn More

Boron tribromide, 99% min

Catalog No. AA3584618
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3584618 50 g
AA3584630 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3584618 Supplier Thermo Scientific Chemicals Supplier No. 03584618

As a catalyst in the manufacturing of diborane, ultra pure boron | CAS: 10294-33-4 | BBr3 | 250.52 g/mol

As a catalyst in the manufacturing of diborane, ultra pure boronBoron tribromide is used as a reactant for luminescent polystyrene derivatives with sterically protected carbazolylborane moieties, high-quality boron-doped graphene via

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10294-33-4
Molecular Formula BBr3
Molecular Weight (g/mol) 250.52
MDL Number MFCD00011312
InChI Key ILAHWRKJUDSMFH-UHFFFAOYSA-N
Synonym boron tribromide, borane, tribromo, boron bromide, tribromoboron, hsdb 327, tribromoboran, boron tribrornide, boron tri bromide, boron tri-bromide
PubChem CID 25134
IUPAC Name tribromoborane
SMILES BrB(Br)Br

Specifications

Boiling Point 90°C
Melting Point -46°C
Color Colorless to Orange
Physical Form Liquid
Quantity 50 g
UN Number UN2692
Merck Index 14,1347
Solubility Information Miscible with ethanol and carbon tetrachloride.
Formula Weight 250.52
Percent Purity ≤99%
Sensitivity Moisture sensitive
Density 2.643 g/mL
Chemical Name or Material Boron tribromide
Show More Show Less
Hazard Category H300+H330-H314-H335
Hazard Statement GHS H Statement
H300-H330-H314
Fatal if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: I; Proper Shipping Name: BORON TRIBROMIDE
EINECSNumber 233-657-9
RTECSNumber ED7400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only