missing translation for 'onlineSavingsMsg'
Learn More

Boron tribromide, 1M solution in methylene chloride, AcroSeal™, Thermo Scientific™

Catalog No. AC427108000
Change view
Click to view available options
800mL; AcroSeal Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC427108000 800mL; AcroSeal Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC427108000 Supplier Thermo Scientific Chemicals Supplier No. 427108000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Lewis acid

Specifications

Chemical Name or Material Boron tribromide
CAS 10294-33-4, 75-09-2
Quantity 800mL
Molecular Formula BBr3
Linear Formula BBr3
MDL Number MFCD00011312
Merck Index 15,1349
Synonym boron tribromide, borane, tribromo, boron bromide, tribromoboron, hsdb 327, tribromoboran, boron tribrornide, boron tri bromide, boron tri-bromide
Solubility Information Solubility in water: reacts. Other solubilities: decomposes in alcohol,soluble in alcohol and ccl4
InChI Key ILAHWRKJUDSMFH-UHFFFAOYSA-N
SMILES BrB(Br)Br
IUPAC Name tribromoborane
Molecular Weight (g/mol) 250.52
PubChem CID 25134
Formula Weight 250.52
Packaging AcroSeal™ Glass bottle
Specific Gravity 1.467
Density 1.4670g/mL
Show More Show Less

RUO – Research Use Only