missing translation for 'onlineSavingsMsg'
Learn More

Boron tribromide, 1M solution in heptane, AcroSeal™

CAS: 10294-33-4 | BBr3 | 250.52 g/mol

$218.32 - $218.32

Specifications

Boiling Point 90.0°C
Chemical Name or Material Boron tribromide
CAS 10294-33-4,142-82-5
Molecular Formula BBr3
MDL Number MFCD00011312
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC378191000
View Documents
Thermo Scientific Chemicals
378191000
100 mL N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

90.0°C
10294-33-4,142-82-5
MFCD00011312
15,1349
Solubility in water: reacts
BrB(Br)Br
250.52
250.52
0.869
Boron tribromide
BBr3
−1°C
boron tribromide, borane, tribromo, boron bromide, tribromoboron, hsdb 327, tribromoboran, boron tribrornide, boron tri bromide, boron tri-bromide
ILAHWRKJUDSMFH-UHFFFAOYSA-N
tribromoborane
25134
AcroSeal™ Glass bottle
0.8690g/mL
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only