Learn More
Boron pieces, crystalline, 20mm (0.8in) & down, 99.5% (metals basis)
CAS: 7440-42-8 | B | 10.81 g/mol
Supplier: Thermo Scientific Chemicals 01213418
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Boron pieces | |
| 50 g | |
| 20mm (0.8 in.) & down | |
| (metals basis) | |
| MFCD00134034 MFCD00151272 | |
| ZOXJGFHDIHLPTG-UHFFFAOYSA-N | |
| boron | |
| 5462311 | |
| 99.5% |
| 7440-42-8 | |
| Odorless | |
| Crystalline Pieces | |
| B | |
| boron, metallic, unii-n9e3x5056q, boron, amorphous, powder, crystalline, boro, boron-elemental, boron, elemental, and compounds, boracium, boranylidyne | |
| [B] | |
| 10.81 | |
| CHEBI:27560 |
Chemical Identifiers
| 7440-42-8 | |
| 10.81 | |
| ZOXJGFHDIHLPTG-UHFFFAOYSA-N | |
| 5462311 | |
| boron |
| B | |
| MFCD00134034 MFCD00151272 | |
| boron, metallic, unii-n9e3x5056q, boron, amorphous, powder, crystalline, boro, boron-elemental, boron, elemental, and compounds, boracium, boranylidyne | |
| CHEBI:27560 | |
| [B] |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 231-151-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only