missing translation for 'onlineSavingsMsg'
Learn More

Boron nitride

Catalog No. AA4060704
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4060704 2 g
AA4060709 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4060704 Supplier Thermo Scientific Chemicals Supplier No. 04060704

CAS: 10043-11-5 | BN | 24.82 g/mol

Boron nitride is used as polymorph, cosmetics, paints, dental cements, and pencil leads. It is also used in xerographic process and laser printers as a charge leakage barrier layer of the photo drum, fillers, processing aids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10043-11-5
Molecular Formula BN
Molecular Weight (g/mol) 24.82
MDL Number MFCD00011317
InChI Key LNLSXDSWJBUPHM-UHFFFAOYSA-N
Synonym boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r
PubChem CID 66227
ChEBI CHEBI:50883
IUPAC Name boranimine
SMILES B=N

Specifications

Color White
Physical Form Powder
Quantity 2 g
Solubility Information Insoluble in water.
Formula Weight 24.82
Odor Odorless
Density 3.48
Chemical Name or Material Boron nitride
Hazard Category H335
Precautionary Statement P261-P271-P304+P340-P312-P501c
EINECSNumber 233-136-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only