missing translation for 'onlineSavingsMsg'
Learn More

Boron nitride, Thermo Scientific Chemicals

Catalog No. AA4060804
Change view
Click to view available options
Quantity:
2 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4060804 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4060804 Supplier Thermo Scientific Chemicals Supplier No. 04060804

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10043-11-5 | Molecular Formula: BN | Molecular Weight: 24.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10043-11-5
Molecular Formula BN
Molecular Weight (g/mol) 24.82
MDL Number MFCD00011317
InChI Key LNLSXDSWJBUPHM-UHFFFAOYSA-N
Synonym boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r
PubChem CID 66227
ChEBI CHEBI:50883
SMILES B=N

Specifications

Physical Form Powder
Quantity 2 g
Odor Odorless
Density 3.48 g/mL
Merck Index 14,1346
IUPAC Name azanylidyneborane
Formula Weight 24.82
Chemical Name or Material Boron nitride
Hazard Category H335
Hazard Statement GHS H Statement
H319-H335
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P271-P304+P340-P312-P501c
EINECSNumber 233-136-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only