missing translation for 'onlineSavingsMsg'
Learn More

Boron Nitride Rod;Diameter (mm), 12.7;Length (mm), 300

Catalog No. AA45912KS
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA45912KS 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA45912KS Supplier Thermo Scientific Chemicals Supplier No. 045912KS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10043-11-5 | BN | 24.82 g/mol

Boron Nitride rod is used as a container for most molten metals like aluminum, iron, steel, silicon, tin germanium and copper due to its properties like corrosion resistance, electrical resistance and the fact that it can withstand temperatures of about 20000. It plays an important role in nanotechnology to produce nanotubes, which find applications in the aerospace industry. It is widely utilized in evaporator boats, nozzles for nonferrous metals, electric insulators in vacuum system, induction heating coil supports, crucibles and radar components and antenna windows and insulators for high temperature furnace.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10043-11-5
Molecular Formula BN
Molecular Weight (g/mol) 24.82
MDL Number MFCD00011317
InChI Key LNLSXDSWJBUPHM-UHFFFAOYSA-N
Synonym boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r
PubChem CID 66227
ChEBI CHEBI:50883
IUPAC Name boranimine
SMILES B=N

Specifications

Quantity 1 Ea.
Odor Odorless
Appearance Solid in various forms
Solubility Information Insoluble in water.
Chemical Name or Material Boron Nitride
Hazard Category H335
Precautionary Statement P261-P271-P304+P340-P312-P501c
EINECSNumber 233-136-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only