missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boron nitride, low binder, 99.5% (metals basis), Thermo Scientific Chemicals
Description
Used as a lubricant and an additive to cosmetic products. Boron nitride has potential use in nanotechnology. Also used in paints, dental cements, pencil leads, xerographic process and laser printers, sealing oxygen sensors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Hot Pressed Rod |
| Quantity | 1 Pc. |
| Packaging | Bag |
| Solubility Information | Insoluble in water. Boron nitride is insoluble in the acids. Soluble in alkaline molten salts and nitrides, |
| Formula Weight | 24.82 |
| Percent Purity | 99.5% |
| Chemical Name or Material | Boron nitride |
Safety and Handling
| Hazard Category | H335 |
| Precautionary Statement | P261-P271-P304+P340-P312-P501c |
| EINECSNumber | 233-136-6 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only