missing translation for 'onlineSavingsMsg'
Learn More

Boron nitride, 99.5% (metals basis), Thermo Scientific™

Catalog No. AA44838KG
Change view
Click to view available options
Quantity:
1 Pc.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA44838KG 1 Pc.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA44838KG Supplier Thermo Scientific Chemicals Supplier No. 44838KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used in production of alloys. It is also used in the manufacture of furnace refractories, insulation materials, machinery and aviation.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 10043-11-5
Molecular Formula BN
Molecular Weight (g/mol) 24.82
MDL Number MFCD00011317
InChI Key LNLSXDSWJBUPHM-UHFFFAOYSA-N
Synonym boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r
PubChem CID 66227
ChEBI CHEBI:50883
IUPAC Name boranylidyneamine
SMILES B=N

Specifications

Physical Form Hot Pressed Rod
Quantity 1 Pc.
Solubility Information Insoluble in water.
Formula Weight 24.82
Percent Purity 99.5%
Packaging Bag
Chemical Name or Material Boron nitride
Hazard Category H335
Precautionary Statement P261-P271-P304+P340-P312-P501c
EINECSNumber 233-136-6
TSCA Yes
Recommended Storage Ambient temperatures