missing translation for 'onlineSavingsMsg'
Learn More

Boron nitride, 98%

Catalog No. AC441802500
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC441802500 250 g
AC441800500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC441802500 Supplier Thermo Scientific Chemicals Supplier No. 441802500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10043-11-5 | BN | 24.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10043-11-5
Molecular Formula BN
Molecular Weight (g/mol) 24.82
MDL Number MFCD00011317
InChI Key LNLSXDSWJBUPHM-UHFFFAOYSA-N
Synonym boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r
PubChem CID 66227
ChEBI CHEBI:50883
SMILES B=N

Specifications

Melting Point 2700.0°C to 3000.0°C
Quantity 250 g
Assay Percent Range 97.5% min
Linear Formula BN
Merck Index 15,1347
Solubility Information Solubility in water: slightly soluble (20°C). Other solubilities: soluble in water (95°)
IUPAC Name azanylidyneborane
Formula Weight 24.82
Percent Purity 98%
Chemical Name or Material Boron nitride
EINECSNumber 233-136-6

RUO – Research Use Only